Merck
  • Efficient clathrin-mediated entry of enteric adenoviruses in human duodenal cells.
Efficient clathrin-mediated entry of enteric adenoviruses in human duodenal cells.
Journal of virology (2023-10-12)
Miriam Becker, Dario Valter Conca, Noemi Dorma, Nitesh Mistry, Elin Hahlin, Lars Frängsmyr, Marta Bally, Niklas Arnberg, Gisa Gerold
ABSTRACT
Enteric adenoviruses have historically been difficult to grow in cell culture, which has resulted in lack of knowledge of host factors and pathways required for infection of these medically relevant viruses. Previous studies in non-intestinal cell lines showed slow infection kinetics and generated comparatively low virus yields compared to other adenovirus types. We suggest duodenum-derived HuTu80 cells as a superior cell line for studies to complement efforts using complex intestinal tissue models. We show that viral host cell factors required for virus entry differ between cell lines from distinct origins and demonstrate the importance of clathrin-mediated endocytosis.
Sigma-Aldrich
Anti-Adenovirus Antibody, clone 20/11, clone 20/11, Chemicon®, from mouse
Sigma-Aldrich
Anti-CAR Antibody, clone RmcB, clone RmcB, Upstate®, from mouse
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone P1E6, azide free, clone P1E6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α3 Antibody, clone ASC-1, clone ASC-1, Chemicon®, from mouse