Merck
  • Loss of LCMT1 and biased protein phosphatase 2A heterotrimerization drive prostate cancer progression and therapy resistance.
Loss of LCMT1 and biased protein phosphatase 2A heterotrimerization drive prostate cancer progression and therapy resistance.
Nature communications (2023-08-30)
Reyaz Ur Rasool, Caitlin M O'Connor, Chandan Kanta Das, Mohammed Alhusayan, Brijesh Kumar Verma, Sehbanul Islam, Ingrid E Frohner, Qu Deng, Erick Mitchell-Velasquez, Jaya Sangodkar, Aqila Ahmed, Sarah Linauer, Ingrid Mudrak, Jessica Rainey, Kaitlin P Zawacki, Tahra K Suhan, Catherine G Callahan, Ryan Rebernick, Ramakrishnan Natesan, Javed Siddiqui, Guido Sauter, Dafydd Thomas, Shaomeng Wang, Derek J Taylor, Ronald Simon, Marcin Cieslik, Arul M Chinnaiyan, Luca Busino, Egon Ogris, Goutham Narla, Irfan A Asangani
ABSTRACT
Loss of the tumor suppressive activity of the protein phosphatase 2A (PP2A) is associated with cancer, but the underlying molecular mechanisms are unclear. PP2A holoenzyme comprises a heterodimeric core, a scaffolding A subunit and a catalytic C subunit, and one of over 20 distinct substrate-directing regulatory B subunits. Methylation of the C subunit regulates PP2A heterotrimerization, affecting B subunit binding and substrate specificity. Here, we report that the leucine carboxy methyltransferase (LCMT1), which methylates the L309 residue of the C subunit, acts as a suppressor of androgen receptor (AR) addicted prostate cancer (PCa). Decreased methyl-PP2A-C levels in prostate tumors is associated with biochemical recurrence and metastasis. Silencing LCMT1 increases AR activity and promotes castration-resistant prostate cancer growth. LCMT1-dependent methyl-sensitive AB56αCme heterotrimers target AR and its critical coactivator MED1 for dephosphorylation, resulting in the eviction of the AR-MED1 complex from chromatin and loss of target gene expression. Mechanistically, LCMT1 is regulated by S6K1-mediated phosphorylation-induced degradation requiring the β-TRCP, leading to acquired resistance to anti-androgens. Finally, feedforward stabilization of LCMT1 by small molecule activator of phosphatase (SMAP) results in attenuation of AR-signaling and tumor growth inhibition in anti-androgen refractory PCa. These findings highlight methyl-PP2A-C as a prognostic marker and that the loss of LCMT1 is a major determinant in AR-addicted PCa, suggesting therapeutic potential for AR degraders or PP2A modulators in prostate cancer treatment.
Sigma-Aldrich
mTOR Inhibitor XI, Torin1, mTOR Inhibitor XI, Torin1, CAS 1222998-36-8, is a cell-permeable, highly potent, ATP-competitive inhibitor of mTOR and DNA-PK (IC50 = 4.32 and 6.34 nM, respectively).
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
NAE Inhibitor, MLN4924
Supelco
Rapamycin, VETRANAL®, analytical standard