Merck
Direct type I interferon signaling in hepatocytes controls malaria.
Cell reports (2022-07-21)
Camila Marques-da-Silva, Kristen Peissig, Michael P Walker, Justine Shiau, Carson Bowers, Dennis E Kyle, Rahul Vijay, Scott E Lindner, Samarchith P Kurup
ABSTRACT
Malaria is a devastating disease impacting over half of the world's population. Plasmodium parasites that cause malaria undergo obligatory development and replication in hepatocytes before infecting red blood cells and initiating clinical disease. While type I interferons (IFNs) are known to facilitate innate immune control to Plasmodium in the liver, how they do so has remained unresolved, precluding the manipulation of such responses to combat malaria. Utilizing transcriptomics, infection studies, and a transgenic Plasmodium strain that exports and traffics Cre recombinase, we show that direct type I IFN signaling in Plasmodium-infected hepatocytes is necessary to control malaria. We also show that the majority of infected hepatocytes naturally eliminate Plasmodium infection, revealing the potential existence of anti-malarial cell-autonomous immune responses in such hepatocytes. These discoveries challenge the existing paradigms in Plasmodium immunobiology and are expected to inspire anti-malarial drugs and vaccine strategies.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-IRF-9 Antibody, clone 6F1-H5, clone 6F1-H5, from mouse