Merck
  • Delphinidin modulates JAK/STAT3 and MAPKinase signaling to induce apoptosis in HCT116 cells.
Delphinidin modulates JAK/STAT3 and MAPKinase signaling to induce apoptosis in HCT116 cells.
Environmental toxicology (2021-05-07)
Zhiyong Zhang, Yan Pan, Yan Zhao, Mudan Ren, Yarui Li, Guifang Lu, Kaichun Wu, Shuixiang He
ABSTRACT
Delphinidin is an anthocyanin that belongs to the group of flavonoids that exert numerous biological activities. However, the molecular mechanisms underlying the anticancer effects of delphinidin remain poorly understood. In our study we analyzed delphinidin modulate STAT-3 and MAPKinase signaling thereby inhbits cell proliferation and promote apoptosis. Our study demonstrated that delphinidin treatment significantly reduced the viability of human colon cancer HCT116 in a concentration-dependent manner. We noticed that delphinidin effectively induced oxidative stress-mediated apoptosis by generating intracellular ROS, decreasing antioxidant levels, inducing lipid peroxidation, and single-strand break on colon cancer cells. In this study, we observed that delphinidin treatment alters the mitochondrial membrane potential, thereby induces apoptosis was closely associated with the induction of pro-apoptotic Bax, Caspase- 3,8 & 9, cytochrome C, and inhibition of anti-apoptotic protein expression. Studies on STAT-3 and MAPKinase signaling showed delphinidin inhibited the phosphorylation of these transcription factors' activity. Inhibition of STAT-3, p38, and ERK1/2 phosphorylation and modulation pro-apoptotic protein expression might be responsible for the anticancer activity of delphinidin in colon cancer cells.
Sigma-Aldrich
Monoclonal Anti-BAD antibody produced in mouse, clone 3H8, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Caspase 8 antibody, Mouse monoclonal, clone CAS8, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Caspase 3 antibody produced in mouse, clone 84803.111, purified immunoglobulin, lyophilized powder
Sigma-Aldrich
Anti-JAK2 antibody, Mouse monoclonal, clone 10.1.5, purified from hybridoma cell culture
Sigma-Aldrich
Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-JAK2 (pTyr1007) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-BAX antibody produced in mouse, clone 1F5-1B7, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-STAT3 (pSer727) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Erk1/2 (Thr202/Tyr204, Thr185/Tyr187)Antibody, recombinant clone AW39R, rabbit monoclonal, clone AW39R, Upstate®, from rabbit
Sigma-Aldrich
Anti-Caspase 9 antibody,Mouse monoclonal, clone CAS9, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Cytochrome C Antibody, Chemicon®, from sheep