Merck
  • Effects of tris (2-carboxyethyl) phosphine hydrochloride treatment on porcine oocyte in vitro maturation and subsequent in vitro fertilized embryo developmental capacity.
Effects of tris (2-carboxyethyl) phosphine hydrochloride treatment on porcine oocyte in vitro maturation and subsequent in vitro fertilized embryo developmental capacity.
Theriogenology (2021-01-15)
Yiren Zeng, Kohei Shinada, Kazuki Hano, Lumin Sui, Ting Yang, Xiangping Li, Takehiro Himaki
ABSTRACT
Oocyte in vitro maturation (IVM) is a crucial process that determines subsequent in vitro embryo production. The present study investigated the effects of the antioxidant tris (2-carboxyethyl) phosphine hydrochloride (TCEP-HCL) on the in vitro maturation of porcine oocytes and in vitro developmental competence of fertilized embryos. Oocytes were matured in IVM medium based on four concentration groups of TCEP-HCL (0, 50, 100, and 200 μM) treatment. 100 μM TCEP-HCL treatment significantly increased the oocyte first polar body extrusion rate, monospermy rate and subsequent in vitro fertilized embryo developmental capacity (cleavage rate, blastocyst formation rate, and blastocyst total cell number) compared to those in the control group. Furthermore, 100 μM TCEP-HCL treatment significantly reduced the levels of reactive oxygen species, significantly increased glutathione levels and mitochondrial content compared to those in the control group. Moreover, 100 μM TCEP-HCL treatment significantly decreased the oocyte apoptosis, blastocyst apoptosis compared to that in the controls. In summary, these results indicate that 100 μM TCEP-HCL treatment improves the quality and developmental capacity of in vitro-fertilized embryos by decreasing oxidative stress in porcine oocytes.
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, suitable for electrophoresis, SDS-PAGE tested
Pricing and availability is not currently available.