Merck
  • Cooperative signaling between oncostatin M, hepatocyte growth factor and transforming growth factor-β enhances epithelial to mesenchymal transition in lung and pancreatic tumor models.
Cooperative signaling between oncostatin M, hepatocyte growth factor and transforming growth factor-β enhances epithelial to mesenchymal transition in lung and pancreatic tumor models.
Cells, tissues, organs (2010-11-03)
Gretchen M Argast, Peter Mercado, Iain J Mulford, Matthew O'Connor, David M Keane, Salam Shaaban, David M Epstein, Jonathan A Pachter, Julie L C Kan
ABSTRACT
Epithelial to mesenchymal transition (EMT) plays a dual role in tumor progression. It enhances metastasis of tumor cells by increasing invasive capacity and promoting survival, and it decreases tumor cell sensitivity to epithelial cell-targeting agents such as epithelial growth factor receptor kinase inhibitors. In order to study EMT in tumor cells, we have characterized 3 new models of ligand-driven EMT: the CFPAC1 pancreatic tumor model and the H358 and H1650 lung tumor models. We identified a diverse set of ligands that drives EMT in these models. Hepatocyte growth factor and oncostatin M induced EMT in all models, while transforming growth factor-β induced EMT in both lung models. We observed morphologic, marker and phenotypic changes in response to chronic ligand treatment. Interestingly, stimulation with 2 ligands resulted in more pronounced EMT compared with single-ligand treatment, demonstrating a spectrum of EMT states induced by parallel signaling, such as the JAK and PI3K pathways. The EMT changes observed in response to the ligand were reversed upon ligand withdrawal, demonstrating the 'metastable' nature of these models. To study the impact of EMT on cell morphology and invasion in a 3D setting, we cultured cells in a semisolid basement membrane extract. Upon stimulation with EMT ligands, the colonies exhibited changes to EMT markers and showed phenotypes ranging from modest differences in colony architecture (CFPAC1) to complex branching structures (H358, H1650). Collectively, these 3 models offer robust cell systems with which to study the roles that EMT plays in cancer progression.
Sigma-Aldrich
MEK Inhibitor I, The MEK Inhibitor I, also referenced under CAS 297744-42-4, controls the biological activity of MEK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 1, DMSO solution
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Millipore
JAK Inhibitor I, JAK Inhibitor I, CAS 457081-03-7, is a potent, reversible, cell-permeable, and ATP-competitive inhibitor of JAK 1 (IC₅₀ = 15 nM), JAK2 (IC₅₀ = 1 nM), JAK3 (Ki = 5 nM) and Tyk2 (IC₅₀ = 1 nM).
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC₅₀ = 1.4 µM). Acts on the ATP-binding site.