Merck
  • Simultaneous determination of spirodiclofen, spiromesifen, and spirotetramat and their relevant metabolites in edible fungi using ultra-performance liquid chromatography/tandem mass spectrometry.
Simultaneous determination of spirodiclofen, spiromesifen, and spirotetramat and their relevant metabolites in edible fungi using ultra-performance liquid chromatography/tandem mass spectrometry.
Scientific reports (2021-01-17)
Fajun Tian, Chengkui Qiao, Caixia Wang, Jing Luo, Linlin Guo, Tao Pang, Jun Li, Ruiping Wang, Rongli Pang, Hanzhong Xie
ABSTRACT
A fast, sensitive, and reliable analytical method was developed and validated for simultaneous identification and quantification of spirodiclofen, spiromesifen, and spirotetramat and their relevant metabolites in edible fungi by ultra-performance liquid chromatography/tandem mass spectrometry (UHPLC-MS/MS). First, sample extraction was done with acetonitrile containing 1% formic acid followed by phase separation with the addition of MgSO4:NaOAc. Then, the supernatant was purified by primary secondary amine (PSA), octadecylsilane (C18), and graphitized carbon black (GCB). The linearities of the calibrations for all analytes were excellent (R2 ≥ 0.9953). Acceptable recoveries (74.5-106.4%) for all analytes were obtained with good intra- and inter- relative standard deviations of less than 14.5%. The limit of quantification (LOQs) for all analytes was 10 μg kg-1. For accurate quantification, matrix-matched calibration curve was applied to normalize the matrix effect. The results indicated that the method was suitable for detecting the three acaricides and their relevant metabolites in edible fungi.
Product Description
Supelco
Spirodiclofen, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Octadecylsilane, 97%