Merck
  • Characterizing ubiquitination sites by peptide-based immunoaffinity enrichment.
Characterizing ubiquitination sites by peptide-based immunoaffinity enrichment.
Molecular & cellular proteomics : MCP (2012-06-26)
Daisy Bustos, Corey E Bakalarski, Yanling Yang, Junmin Peng, Donald S Kirkpatrick
ABSTRACT
Advances in high resolution tandem mass spectrometry and peptide enrichment technologies have transformed the field of protein biochemistry by enabling analysis of end points that have traditionally been inaccessible to molecular and biochemical techniques. One field benefitting from this research has been the study of ubiquitin, a 76-amino acid protein that functions as a covalent modifier of other proteins. Seminal work performed decades ago revealed that trypsin digestion of a branched protein structure known as A24 yielded an enigmatic diglycine signature bound to a lysine residue in histone 2A. With the onset of mass spectrometry proteomics, identification of K-GG-modified peptides has emerged as an effective way to map the position of ubiquitin modifications on a protein of interest and to quantify the extent of substrate ubiquitination. The initial identification of K-GG peptides by mass spectrometry initiated a flurry of work aimed at enriching these post-translationally modified peptides for identification and quantification en masse. Recently, immunoaffinity reagents have been reported that are capable of capturing K-GG peptides from ubiquitin and its thousands of cellular substrates. Here we focus on the history of K-GG peptides, their identification by mass spectrometry, and the utility of immunoaffinity reagents for studying the mechanisms of cellular regulation by ubiquitin.
Sigma-Aldrich
Gly-Gly, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Gly-Gly, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Gly-Gly, BioPerformance Certified, suitable for cell culture, ≥99%
Sigma-Aldrich
Gly-Gly, ≥99% (titration)
Sigma-Aldrich
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid