Merck
  • TGF-β-mediated silencing of genomic organizer SATB1 promotes Tfh cell differentiation and formation of intra-tumoral tertiary lymphoid structures.
TGF-β-mediated silencing of genomic organizer SATB1 promotes Tfh cell differentiation and formation of intra-tumoral tertiary lymphoid structures.
Immunity (2022-01-13)
Ricardo A Chaurio, Carmen M Anadon, Tara Lee Costich, Kyle K Payne, Subir Biswas, Carly M Harro, Carlos Moran, Antonio C Ortiz, Carla Cortina, Kristen E Rigolizzo, Kimberly B Sprenger, Jessica A Mine, Patrick Innamarato, Gunjan Mandal, John J Powers, Alexandra Martin, Zhitao Wang, Sumit Mehta, Bradford A Perez, Roger Li, John Robinson, Jodi L Kroeger, Tyler J Curiel, Xiaoqing Yu, Paulo C Rodriguez, Jose R Conejo-Garcia
ABSTRACT
The immune checkpoint receptor PD-1 on T follicular helper (Tfh) cells promotes Tfh:B cell interactions and appropriate positioning within tissues. Here, we examined the impact of regulation of PD-1 expression by the genomic organizer SATB1 on Tfh cell differentiation. Vaccination of CD4CreSatb1f/f mice enriched for antigen-specific Tfh cells, and TGF-β-mediated repression of SATB1 enhanced Tfh differentiation of human T cells. Mechanistically, high Icos expression in Satb1-/- CD4+ T cells promoted Tfh cell differentiation by preventing T follicular regulatory cell skewing and resulted in increased isotype-switched B cell responses in vivo. Ovarian tumors in CD4CreSatb1f/f mice accumulated tumor antigen-specific, LIGHT+CXCL13+IL-21+ Tfh cells and tertiary lymphoid structures (TLS). TLS formation decreased tumor growth in a CD4+ T cell and CXCL13-dependent manner. The transfer of Tfh cells, but not naive CD4+ T cells, induced TLS at tumor beds and decreased tumor growth. Thus, TGF-β-mediated silencing of Satb1 licenses Tfh cell differentiation, providing insight into the genesis of TLS within tumors.
Sigma-Aldrich
Anti-Peripheral Node Addressin (PNAd) Antibody, clone MECA-79, clone MECA-79, from rat
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)