Merck
  • Three-dimensional superresolution fluorescence microscopy maps the variable molecular architecture of the nuclear pore complex.
Three-dimensional superresolution fluorescence microscopy maps the variable molecular architecture of the nuclear pore complex.
Molecular biology of the cell (2021-07-01)
Vilma Jimenez Sabinina, M Julius Hossain, Jean-Karim Hériché, Philipp Hoess, Bianca Nijmeijer, Shyamal Mosalaganti, Moritz Kueblbeck, Andrea Callegari, Anna Szymborska, Martin Beck, Jonas Ries, Jan Ellenberg
ABSTRACT
Nuclear pore complexes (NPCs) are large macromolecular machines that mediate the traffic between the nucleus and the cytoplasm. In vertebrates, each NPC consists of ∼1000 proteins, termed nucleoporins, and has a mass of more than 100 MDa. While a pseudo-atomic static model of the central scaffold of the NPC has recently been assembled by integrating data from isolated proteins and complexes, many structural components still remain elusive due to the enormous size and flexibility of the NPC. Here, we explored the power of three-dimensional (3D) superresolution microscopy combined with computational classification and averaging to explore the 3D structure of the NPC in single human cells. We show that this approach can build the first integrated 3D structural map containing both central as well as peripheral NPC subunits with molecular specificity and nanoscale resolution. Our unbiased classification of more than 10,000 individual NPCs indicates that the nuclear ring and the nuclear basket can adopt different conformations. Our approach opens up the exciting possibility to relate different structural states of the NPC to function in situ.
Sigma-Aldrich
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF680 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Anti-AHCTF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution