Merck
  • Thyroid hormone signaling promotes hepatic lipogenesis through the transcription factor ChREBP.
Thyroid hormone signaling promotes hepatic lipogenesis through the transcription factor ChREBP.
Science signaling (2021-11-17)
Arturo Mendoza, Catherine Tang, Jinyoung Choi, Mariana Acuña, Maya Logan, Adriana G Martin, Lujain Al-Sowaimel, Bhavna N Desai, Danielle E Tenen, Christopher Jacobs, Anna Lyubetskaya, Yulong Fu, Hong Liu, Linus Tsai, David E Cohen, Douglas Forrest, Andrew A Wilson, Anthony N Hollenberg
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-SREBP-1 Antibody, clone 20B12, clone 20B12, from rabbit
Sigma-Aldrich
Anti-Actin (20-33) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-HDAC1 Antibody, clone 2E10, clone 2E10, Upstate®, from mouse