Merck
  • TRPM4 mediates a subthreshold membrane potential oscillation in respiratory chemoreceptor neurons that drives pacemaker firing and breathing.
TRPM4 mediates a subthreshold membrane potential oscillation in respiratory chemoreceptor neurons that drives pacemaker firing and breathing.
Cell reports (2021-02-04)
Keyong Li, Stephen B G Abbott, Yingtang Shi, Pierce Eggan, Elizabeth C Gonye, Douglas A Bayliss
ABSTRACT
Brainstem networks that control regular tidal breathing depend on excitatory drive, including from tonically active, CO2/H+-sensitive neurons of the retrotrapezoid nucleus (RTN). Here, we examine intrinsic ionic mechanisms underlying the metronomic firing activity characteristic of RTN neurons. In mouse brainstem slices, large-amplitude membrane potential oscillations are evident in synaptically isolated RTN neurons after blocking action potentials. The voltage-dependent oscillations are abolished by sodium replacement; blocking calcium channels (primarily L-type); chelating intracellular Ca2+; and inhibiting TRPM4, a Ca2+-dependent cationic channel. Likewise, oscillation voltage waveform currents are sensitive to calcium and TRPM4 channel blockers. Extracellular acidification and serotonin (5-HT) evoke membrane depolarization that augments TRPM4-dependent oscillatory activity and action potential discharge. Finally, inhibition of TRPM4 channels in the RTN of anesthetized mice reduces central respiratory output. These data implicate TRPM4 in a subthreshold oscillation that supports the pacemaker-like firing of RTN neurons required for basal, CO2-stimulated, and state-dependent breathing.
Sigma-Aldrich
9-Phenanthrol, technical grade
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Supelco
Flufenamic acid, analytical standard, for drug analysis
Sigma-Aldrich
Tetraethylammonium chloride, ≥98% (titration)
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
Strychnine hydrochloride
Sigma-Aldrich
4-Nitrophenyl β-D-lactopyranoside, powder
Sigma-Aldrich
ML252, ≥98% (HPLC)
Sigma-Aldrich
(+)-Bicuculline, ≥97.0% (TLC)
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat