Merck
  • Efficient Luminescent Halide Quadruple-Perovskite Nanocrystals via Trap-Engineering for Highly Sensitive Photodetectors.
Efficient Luminescent Halide Quadruple-Perovskite Nanocrystals via Trap-Engineering for Highly Sensitive Photodetectors.
Advanced materials (Deerfield Beach, Fla.) (2021-01-21)
Tianxin Bai, Bin Yang, Junsheng Chen, Daoyuan Zheng, Zhe Tang, Xiaochen Wang, Yang Zhao, Ruifeng Lu, Keli Han
ABSTRACT
The colloidal synthesis of a new type of lead-free halide quadruple-perovskite nanocrystals (NCs) is reported. The photoluminescence quantum yield and charge-carrier lifetime of quadruple-perovskite NCs can be enhanced by 96 and 77-fold, respectively, via metal alloying. Study of charge-carrier dynamics provide solid demonstrate that the PL enhancement is due to the elimination of ultrafast (1.4 ps) charge-carrier trapping processes in the alloyed NCs. Thanks to the high crystallinity, low trap-state density, and long carrier lifetime (193.4 μs), the alloyed quadruple-perovskite NCs can serve as the active material for high-performance photodetectors, which exhibit high responsivity (up to 0.98 × 104 A W-1 ) and an external quantum efficiency (EQE) of 3 × 106 %. These numbers are among the highest for perovskite-NC-based photodetectors.
Sigma-Aldrich
Antimony(III) acetate, 99.99% trace metals basis
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Manganese(II) acetate, 98%
Sigma-Aldrich
Cesium acetate, 99.9% trace metals basis
Sigma-Aldrich
Bismuth(III) acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%