Merck
  • Immortalized keratinocytes cells generates an effective model of Epidermal Human Equivalent for irritation and corrosion tests.
Immortalized keratinocytes cells generates an effective model of Epidermal Human Equivalent for irritation and corrosion tests.
Toxicology in vitro : an international journal published in association with BIBRA (2020-12-15)
C A Mini, S A C Dreossi, F R Abe, S S Maria-Engler, D P Oliveira
ABSTRACT
Three-dimensional skin models, also named 3D skin models, human skin equivalents (HSEs), or Human Epidermal Equivalents (HEEs), have been increasingly used for chemical assessments in terms of efficacy and safety. Considering this, we developed an HEE model using immortalized HaCaT cells, aiming to overcome the limitation of primary tissue source. Our 3D model (HaCaT-HEE) exhibited important markers of cell differentiation (CK10, CK14, involucrin, and filaggrin), although the stratum corneum was shown to be modest. Besides, the model showed a good prediction potential considering membrane permeability, sensitivity, specificity, and accuracy in distinguishing irritant and corrosive effects after exposure to selected chemicals recommended by the OECD protocols. We also validated the formazan determination for the MTT method using High-Performance Liquid Chromatography (HPLC). For that, we considered carry over, linearity, reproducibility/robustness, accuracy, precision, selectivity, and matrix effect, according to the Food and Drug Administration (FDA) guideline. Based on our results, we can conclude that our model has an acceptable predictive value for the safety evaluation of compounds after skin exposure, with the great advantage of being constructed using immortalized cells.
Sigma-Aldrich
Heptyl butyrate, ≥98%, FG
Sigma-Aldrich
2-Methyl-3-(p-isopropylphenyl)propionaldehyde, ≥95%, FCC, FG
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
apo-Transferrin human, powder, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
1-Methyl-3-phenylpiperazine, 97%
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone 21-hemisuccinate sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)