Merck
Generation of Functional Mouse Hippocampal Neurons.
Bio-protocol (2020-09-29)
Francesco Tomassoni-Ardori, Zhenyi Hong, Gianluca Fulgenzi, Lino Tessarollo
ABSTRACT
Primary culture of mouse hippocampal neurons is a very useful in vitro model for studying neuronal development, axonal and dendritic morphology, synaptic functions, and many other neuronal features. Here we describe a step-by-step process of generating primary neurons from mouse embryonic hippocampi (E17.5/E18.5). Hippocampal neurons generated with this protocol can be plated in different tissue culture dishes according to different experimental aims and can produce a reliable source of pure and differentiated neurons in less than one week. This protocol covers all the steps necessary for the preparation, culture and characterization of the neuronal culture, including the illustration of dissection instruments, surgical procedure for embryos' isolation, culturing conditions and assessment of culture's purity and differentiation. Evaluation of neuronal activity was performed by analysis of calcium imaging dynamics at six days in culture.
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Donkey Anti-Rabbit IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Donkey Anti-Mouse IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Anti-Fox1 Antibody, clone 1D10, clone 1D10, from mouse
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse