Merck
  • In vivo evaluation of diaminodiphenyls: anticonvulsant agents with minimal acute neurotoxicity.
In vivo evaluation of diaminodiphenyls: anticonvulsant agents with minimal acute neurotoxicity.
Bioorganic & medicinal chemistry letters (2009-07-28)
David R Worthen, Aimee K Bence, James P Stables, Peter A Crooks
ABSTRACT
Several diaminodiphenyl analogs were assessed in vivo for their capacity to inhibit seizure induction and propagation in rodents. Both 3,4'- and 4,4'-diaminodiphenyl compounds prevented seizures for as long as 4h after maximal electric shock induction. 4,4'-Diphenyl compounds bridged by a methylene, sulfide, or carbonyl linker also attenuated focal seizure acquisition in a kindling model. Of these analogs, based upon data generated in two rodent species, 4,4'-thiodianiline (1) was identified as the most active compound, significantly reducing seizure staging scores and after-discharge duration for several hours after systemic administration. All compounds were devoid of acute in vivo neurotoxicity at doses well above those required for anticonvulsant activity.
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Supelco
4,4′-Diaminodiphenylmethane, analytical standard
Sigma-Aldrich
4,4′-Diaminodiphenyl sulfide, 98%
Sigma-Aldrich
4,4′-Oxydianiline, 97%
Supelco
4,4′-Diaminodiphenyl sulfide, analytical standard
Sigma-Aldrich
4,4′-Diaminobenzophenone, 97%