Merck
  • Regulation of Aluminum Resistance in Arabidopsis Involves the SUMOylation of the Zinc Finger Transcription Factor STOP1.
Regulation of Aluminum Resistance in Arabidopsis Involves the SUMOylation of the Zinc Finger Transcription Factor STOP1.
The Plant cell (2020-10-23)
Qiu Fang, Jie Zhang, Yang Zhang, Ni Fan, Harrold A van den Burg, Chao-Feng Huang, Qiu Fang, Jie Zhang, Yang Zhang, Ni Fan, Harrold A van den Burg, Chao-Feng Huang
ABSTRACT
Aluminum (Al) is a primary constraint for crop production on acid soils, which make up more than 30% of the arable land in the world. Al resistance in Arabidopsis (Arabidopsis thaliana) is achieved by malate secretion mediated by the Al-ACTIVATED MALATE TRANSPORTER1 (AtALMT1) transporter. The C2H2-type transcription factor SENSITIVE TO PROTON RHIZOTOXICITY1 (STOP1) is essential and required for Al resistance, where it acts by inducing the expression of Al-resistance genes, including AtALMT1 In this study, we report that STOP1 protein function is modified by SUMOylation. The SMALL UBIQUITIN-LIKE MODIFIER (SUMO) protease ESD4, but not other SUMO proteases, specifically interacts with and deSUMOylates STOP1. Mutation of ESD4 increases the level of STOP1 SUMOylation and the expression of the STOP1-regulated gene AtALMT1, which contributes to the increased Al resistance in esd4 The esd4 mutation does not influence STOP1 protein abundance but increases the association of STOP1 with the AtALMT1 promoter, which might explain the elevated expression of AtALMT1 in esd4 We demonstrate that STOP1 is mono-SUMOylated at K40, K212, or K395 sites, and blocking STOP1 SUMOylation reduces STOP1 stability and the expression of STOP1-regulated genes, leading to the reduced Al resistance. Our results thus reveal the involvement of SUMOylation in the regulation of STOP1 and Al resistance in Arabidopsis.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Gelzan CM, Gelrite®
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Ammonium sulfamate, BioXtra, ≥98.0%