Merck
  • Macrophage development and activation involve coordinated intron retention in key inflammatory regulators.
Macrophage development and activation involve coordinated intron retention in key inflammatory regulators.
Nucleic acids research (2020-05-26)
Immanuel D Green, Natalia Pinello, Renhua Song, Quintin Lee, James M Halstead, Chau-To Kwok, Alex C H Wong, Shalima S Nair, Susan J Clark, Ben Roediger, Ulf Schmitz, Mark Larance, Rippei Hayashi, John E J Rasko, Justin J-L Wong
ABSTRACT
Monocytes and macrophages are essential components of the innate immune system. Herein, we report that intron retention (IR) plays an important role in the development and function of these cells. Using Illumina mRNA sequencing, Nanopore direct cDNA sequencing and proteomics analysis, we identify IR events that affect the expression of key genes/proteins involved in macrophage development and function. We demonstrate that decreased IR in nuclear-detained mRNA is coupled with increased expression of genes encoding regulators of macrophage transcription, phagocytosis and inflammatory signalling, including ID2, IRF7, ENG and LAT. We further show that this dynamic IR program persists during the polarisation of resting macrophages into activated macrophages. In the presence of proinflammatory stimuli, intron-retaining CXCL2 and NFKBIZ transcripts are rapidly spliced, enabling timely expression of these key inflammatory regulators by macrophages. Our study provides novel insights into the molecular factors controlling vital regulators of the innate immune response.
Product Description
Supelco
Empore SPE Disks, matrix active group SDB-RPS, diam. 47 mm, pk of 20
Sigma-Aldrich
Donkey Anti-Mouse IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Donkey Anti-Rabbit IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Anti-DIS3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution