Merck
  • Direct transcriptional control of a p38 MAPK pathway by the circadian clock in Neurospora crassa.
Direct transcriptional control of a p38 MAPK pathway by the circadian clock in Neurospora crassa.
PloS one (2011-11-17)
Teresa M Lamb, Charles S Goldsmith, Lindsay Bennett, Katelyn E Finch, Deborah Bell-Pedersen
ABSTRACT
MAPK signal transduction pathways are important regulators of stress responses, cellular growth, and differentiation. In Neurospora, the circadian clock controls rhythms in phosphorylation of the p38-like MAPK (OS-2); however, the mechanism for this regulation is not known. We show that the WCC, a transcription factor and clock component, binds to the os-4 MAPKKK promoter in response to light and rhythmically in constant darkness, peaking in the subjective morning. Deletion of the WCC binding sites in the os-4 promoter disrupts both os-4 mRNA and OS-2 phosphorylation rhythms. The clock also indirectly regulates rhythmic expression of the histidyl-phosphotransferase gene, hpt-1, which peaks in the evening. Anti-phase expression of positive (OS-4) and negative (HPT-1) MAPK pathway regulators likely coordinate to enhance rhythmic MAPK activation to prepare cells to respond to osmotic stress during the day in the natural environment. Consistent with this idea, we show that wild type cells have a clock-dependent morning kinetic advantage in glycerol accumulation after salt stress as compared to evening treatment. Thus, circadian transcriptional control of MAPK pathway components leads to striking time-of-day-specific effects on the signaling status and physiological response of the pathway.
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Formaldehyde-2,4-dinitrophenylhydrazone, BCR®, certified reference material
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Glycerol Standard Solution, 2.5 mg/ml equivalent triolein concentration
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Formaldehyde in Water