Merck
  • The Zebrafish Dorsolateral Habenula Is Required for Updating Learned Behaviors.
The Zebrafish Dorsolateral Habenula Is Required for Updating Learned Behaviors.
Cell reports (2020-08-28)
Fabrizio Palumbo, Bram Serneels, Robbrecht Pelgrims, Emre Yaksi
ABSTRACT
Operant learning requires multiple cognitive processes, such as learning, prediction of potential outcomes, and decision-making. It is less clear how interactions of these processes lead to the behavioral adaptations that allow animals to cope with a changing environment. We show that juvenile zebrafish can perform conditioned place avoidance learning, with improving performance across development. Ablation of the dorsolateral habenula (dlHb), a brain region involved in associative learning and prediction of outcomes, leads to an unexpected improvement in performance and delayed memory extinction. Interestingly, the control animals exhibit rapid adaptation to a changing learning rule, whereas dlHb-ablated animals fail to adapt. Altogether, our results show that the dlHb plays a central role in switching animals' strategies while integrating new evidence with prior experience.
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Metronidazole, BioXtra