Merck
  • Suspension fluorescence in situ hybridization (S-FISH) combined with automatic detection and laser microdissection for STR profiling of male cells in male/female mixtures.
Suspension fluorescence in situ hybridization (S-FISH) combined with automatic detection and laser microdissection for STR profiling of male cells in male/female mixtures.
International journal of legal medicine (2009-03-26)
Mado Vandewoestyne, David Van Hoofstat, Filip Van Nieuwerburgh, Dieter Deforce
ABSTRACT
Laser microdissection is a valuable tool for isolating specific cells from mixtures, such as male cells in a mixture with female cells, e.g., in cases of sexual assault. These cells can be stained with Y-chromosome-specific probes. We developed an automatic screening method to detect male cells after fluorescence in situ hybridization in suspension (S-FISH). To simulate forensic casework, the method was tested on female saliva after cataglottis (a kiss involving tongue-to-tongue contact) and on licking traces (swabs of dried male saliva on female skin) even after drying. After isolation of the detected cells, short tandem repeat profiling was performed. Full DNA profiles could consistently be obtained from as little as ten buccal cells. Isolation of five cells resulted in a mean of 98% (SD of 3.4%) of the alleles detected, showing that the developed S-FISH staining had no significant negative influence on DNA recovery and can be used in forensic casework.
Sigma-Aldrich
Poly-L-lysine solution, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly-L-lysine solution, mol wt 150,000-300,000, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
Sigma-Aldrich
Poly-L-lysine solution, 0.1 % (w/v) in H2O
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Formamide, JIS special grade, ≥98.5%
Sigma-Aldrich
Formamide, SAJ first grade, ≥98.5%
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%