Merck
  • ERα-dependent stimulation of LCN2 in uterine epithelium during mouse early pregnancy.
ERα-dependent stimulation of LCN2 in uterine epithelium during mouse early pregnancy.
Reproduction (Cambridge, England) (2020-01-23)
Y-F Liu, M-Y Li, Y-P Yan, W Wei, B Li, H-Y Pan, Z-M Yang, X-H Liang
ABSTRACT
Maintenance of a suitable uterine milieu is important for embryo development and subsequent implantation during early pregnancy. High estrogen level in proestrous and estrous stages is essential for uterine anti-bacterial activity during preimplantation period. Lipocalin-2 is an essential molecule which prevents bacterial infection by sequestering iron. In this study, the highest expression of lipocalin-2 is observed in the endometrial epithelium on day 1 of normal pregnancy and pseudopregnancy, which exhibit a similar hormone scenario. By injecting the agonists for estrogen receptor α and estrogen receptor β in ovariectomized mice, we found estrogen receptor α is the dominant member for estrogen regulation on lipocalin-2 expression. Estrogen treatment in estrogen receptor α-knockout mice further confirmed the role of estrogen receptor α. Using published data from whole-genome estrogen receptor α binding site assay, significant estrogen receptor α recruitment peaks are found at the downstream of lipocalin-2 gene after estrogen treatment. Furthermore, to study the anti-bacterial activity of lipocalin-2 in uterus, Escherichia coli is injected to mimic bacterial infection. Our results showed an obvious induction of lipocalin-2 in Escherichia coli-treated group. Taken together, this study indicates estrogen regulation of lipocalin-2 in uterine epithelium is mediated by estrogen receptor α, and lipocalin-2 may have anti-bacterial activity during early pregnancy.
Sigma-Aldrich
1,3,5-Tris(4-hydroxyphenyl)-4-propyl-1H-pyrazole, powder, ≥98% (HPLC)
Sigma-Aldrich
Bovine Serum Albumin, chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Methyl Green, zinc chloride salt, ~85%
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Roche
DIG RNA Labeling Kit (SP6/T7), sufficient for 2 x 10 labeling reactions, kit of 1 (12 components), suitable for hybridization, suitable for Southern blotting
SAFC
Hanks′ Balanced Salts, Modified, without calcium chloride, magnesium sulfate, phenol red and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
2,3-Bis(4-hydroxyphenyl)propionitrile, ≥98% (HPLC)
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate and phenol red, powder, suitable for cell culture
Sigma-Aldrich
Sesame oil, Antioxidant, delivery vehicle for fat-soluble compounds
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
(−)-Tetramisole hydrochloride, ≥99% (GC)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%