Merck
  • E3 Ligase Ligands for PROTACs: How They Were Found and How to Discover New Ones.
E3 Ligase Ligands for PROTACs: How They Were Found and How to Discover New Ones.
SLAS discovery : advancing life sciences R & D (2020-11-05)
Tasuku Ishida, Alessio Ciulli
ABSTRACT
Bifunctional degrader molecules, also called proteolysis-targeting chimeras (PROTACs), are a new modality of chemical tools and potential therapeutics to understand and treat human disease. A required PROTAC component is a ligand binding to an E3 ubiquitin ligase, which is then joined to another ligand binding to a protein to be degraded via the ubiquitin-proteasome system. The advent of nonpeptidic small-molecule E3 ligase ligands, notably for von Hippel-Lindau (VHL) and cereblon (CRBN), revolutionized the field and ushered in the design of drug-like PROTACs with potent and selective degradation activity. A first wave of PROTAC drugs are now undergoing clinical development in cancer, and the field is seeking to extend the repertoire of chemistries that allow hijacking new E3 ligases to improve the scope of targeted protein degradation.Here, we briefly review how traditional E3 ligase ligands were discovered, and then outline approaches and ligands that have been recently used to discover new E3 ligases for PROTACs. We will then take an outlook at current and future strategies undertaken that invoke either target-based screening or phenotypic-based approaches, including the use of DNA-encoded libraries (DELs), display technologies and cyclic peptides, smaller molecular glue degraders, and covalent warhead ligands. These approaches are ripe for expanding the chemical space of PROTACs and usher in the advent of other emerging bifunctional modalities of proximity-based pharmacology.
Sigma-Aldrich
E7820-PEG1-NH2 hydrochloride
Sigma-Aldrich
E7820-C3-NH2 hydrochloride
Sigma-Aldrich
BocA1V1PF2-OC6-NH2 hydrochloride, ≥95%
Sigma-Aldrich
A1V1PF2-OEt-PEG3-NH2 hydrochloride
Sigma-Aldrich
BocA1V2PF1-NHPEG1-NH2
Sigma-Aldrich
A1V1PF2-OEt-PEG1-NH2 hydrochloride
Sigma-Aldrich
A1V2PF1-NHEt-C10-NH2
Sigma-Aldrich
A1V2PF2-NHEt-PEG3-NH2, ≥95%
Sigma-Aldrich
A1V2PF1-NHEt-PEG3-NH2
Sigma-Aldrich
A1V2PF1-NHEt-PEG1-NH2, ≥95%
Sigma-Aldrich
BocA1V2PF2
Sigma-Aldrich
BocA1V2PF2-NHC10-NH2, ≥85%
Sigma-Aldrich
A1V2PF2-NHEt-C6-NH2
Sigma-Aldrich
BocA1V1PF2-OC10-NH2 hydrochloride
Sigma-Aldrich
A1V2PF2-NHEt-C10-NH2, ≥95%
Sigma-Aldrich
A1V2PF1-NHEt-C6-NH2
Sigma-Aldrich
BocA1V2PF2-NHC6-NH2
Sigma-Aldrich
BocA1V2PF2-NHPEG3-NH2, ≥95%
Sigma-Aldrich
A1V1PF2-OEt-C6-NH2 hydrochloride
Sigma-Aldrich
E7820-C9-NH2 hydrochloride
Sigma-Aldrich
A1V1PF2-OEt, ≥95%
Sigma-Aldrich
A1V2PF1-NHEt, ≥95%
Sigma-Aldrich
A1V1PF2-OEt-C10-NH2 hydrochloride
Sigma-Aldrich
A1V2PF2-NHEt, ≥95%
Sigma-Aldrich
A1V2PF2-NHEt-PEG1-NH2
Sigma-Aldrich
BocA1V2PF1-NHC6-NH2
Sigma-Aldrich
BocA1V1PF2-OPEG3-NH2 hydrochloride
Sigma-Aldrich
BocA1V1PF2, ≥95%
Sigma-Aldrich
BocA1V2PF1-NHC10-NH2
Sigma-Aldrich
BocA1V1PF2-OPEG1-NH2 hydrochloride, ≥95%