Merck
  • Recapitulating idiopathic pulmonary fibrosis related alveolar epithelial dysfunction in a human iPSC-derived air-liquid interface model.
Recapitulating idiopathic pulmonary fibrosis related alveolar epithelial dysfunction in a human iPSC-derived air-liquid interface model.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-04-17)
Eva Schruf, Victoria Schroeder, Huy Q Le, Tanja Schönberger, Dagmar Raedel, Emily L Stewart, Katrin Fundel-Clemens, Teresa Bluhmki, Sabine Weigle, Michael Schuler, Matthew J Thomas, Ralf Heilker, Megan J Webster, Martin Dass, Manfred Frick, Birgit Stierstorfer, Karsten Quast, James P Garnett
ABSTRACT
Idiopathic pulmonary fibrosis (IPF) is a fatal disease of unknown cause that is characterized by progressive fibrotic lung remodeling. An abnormal emergence of airway epithelial-like cells within the alveolar compartments of the lung, herein termed bronchiolization, is often observed in IPF. However, the origin of this dysfunctional distal lung epithelium remains unknown due to a lack of suitable human model systems. In this study, we established a human induced pluripotent stem cell (iPSC)-derived air-liquid interface (ALI) model of alveolar epithelial type II (ATII)-like cell differentiation that allows us to investigate alveolar epithelial progenitor cell differentiation in vitro. We treated this system with an IPF-relevant cocktail (IPF-RC) to mimic the pro-fibrotic cytokine milieu present in IPF lungs. Stimulation with IPF-RC during differentiation increases secretion of IPF biomarkers and RNA sequencing (RNA-seq) of these cultures reveals significant overlap with human IPF patient data. IPF-RC treatment further impairs ATII differentiation by driving a shift toward an airway epithelial-like expression signature, providing evidence that a pro-fibrotic cytokine environment can influence the proximo-distal differentiation pattern of human lung epithelial cells. In conclusion, we show for the first time, the establishment of a human model system that recapitulates aspects of IPF-associated bronchiolization of the lung epithelium in vitro.
Sigma-Aldrich
Anti-SOX9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MUC5B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SFTPC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-Prosurfactant Protein C (proSP-C) Antibody, serum, Chemicon®
Sigma-Aldrich
Human MMP-7 ELISA Kit, on for cell culture supernatant
Sigma-Aldrich
Human MMP-10 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Anti-Collagen Type I antibody, Mouse monoclonal, clone COL-1, purified from hybridoma cell culture