Merck
Personalized Mapping of Drug Metabolism by the Human Gut Microbiome.
Cell (2020-06-12)
Bahar Javdan, Jaime G Lopez, Pranatchareeya Chankhamjon, Ying-Chiang J Lee, Raphaella Hull, Qihao Wu, Xiaojuan Wang, Seema Chatterjee, Mohamed S Donia
ABSTRACT
The human gut microbiome harbors hundreds of bacterial species with diverse biochemical capabilities. Dozens of drugs have been shown to be metabolized by single isolates from the gut microbiome, but the extent of this phenomenon is rarely explored in the context of microbial communities. Here, we develop a quantitative experimental framework for mapping the ability of the human gut microbiome to metabolize small molecule drugs: Microbiome-Derived Metabolism (MDM)-Screen. Included are a batch culturing system for sustained growth of subject-specific gut microbial communities, an ex vivo drug metabolism screen, and targeted and untargeted functional metagenomic screens to identify microbiome-encoded genes responsible for specific metabolic events. Our framework identifies novel drug-microbiome interactions that vary between individuals and demonstrates how the gut microbiome might be used in drug development and personalized medicine.
Product Description
Supelco
Capecitabine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Mycophenolate mofetil, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Supelco
Metronidazole, analytical standard
Sigma-Aldrich
Nicardipine hydrochloride, powder, ≥98%
Sigma-Aldrich
Misoprostol, ≥99% (HPLC)
Sigma-Aldrich
5-Fluoro-5′-deoxyuridine
Supelco
Risperidone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Misoprostol free acid, ≥90%
Supelco
Sulfamethoxazole
Supelco
Aspartame, analytical standard
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
Mycophenolic acid, ≥98%
Sigma-Aldrich
Clonazepam, powder
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Sigma-Aldrich
Clofazimine
Supelco
Bisacodyl, analytical standard, for drug analysis
Supelco
Zidovudine, Pharmaceutical Secondary Standard; Certified Reference Material