Merck
  • Effect of the functionalisation agent on the surface-enhanced Raman scattering (SERS) spectrum: Case study of pyridine derivatives.
Effect of the functionalisation agent on the surface-enhanced Raman scattering (SERS) spectrum: Case study of pyridine derivatives.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2020-03-13)
Elodie Dumont, Charlotte De Bleye, Merzouk Haouchine, Laureen Coïc, Pierre-Yves Sacré, Philippe Hubert, Eric Ziemons
ABSTRACT
Nowadays, the use of functionalised surface-enhanced Raman scattering (SERS) substrates has become common. These surface modifying agents notably act as Raman reporters, as sensors of biological processes (pH, redox probes) or to increase the sensitivity and/or the specificity of SERS detections. However, the effects of the functionalisation agents are deeply examined in very few studies, even though they can affect the aggregation behaviour of the SERS substrate. Moreover, depending on their concentration and on the pH, their spectral signature can be modified and they can even degrade if stored inappropriately. In this context, this paper aims at emphasising the importance of the different aspects previously listed in the selection of a functionalisation agent. Pyridine derivatives were picked out to highlight these parameters, as some of these compounds are commonly used to be grafted onto SERS substrates. Two widespread syntheses of nanoparticles were selected as SERS substrates: citrate-reduced gold and silver nanoparticles. The surface of the nanoparticles was functionalised with several pyridine derivatives at different concentrations and in several solvents. It was observed that the molecules under study had a concentration-dependent effect on nanoparticle aggregation. A stability study was furthermore conducted in order to determine the best preservation conditions of the grafting solutions. In conclusion, this paper shines a light on the relevance of the investigation of the too-often neglected behaviour of the surface modifying agents. Before their application in SERS analyses, parameters such as the label concentration should therefore be included in an experimental design to optimise the sample preparation.
Sigma-Aldrich
2-pyridylethylmercaptan, AldrichCPR
Sigma-Aldrich
4-Mercaptopyridine, 95%
Sigma-Aldrich
2-Mercaptopurine, ≥95%, crystalline
Sigma-Aldrich
2-Mercaptopyridine, ReagentPlus®, 99%
Sigma-Aldrich
Thiophenol, ≥99%