Merck
  • A Quantitative Tissue-Specific Landscape of Protein Redox Regulation during Aging.
A Quantitative Tissue-Specific Landscape of Protein Redox Regulation during Aging.
Cell (2020-02-29)
Haopeng Xiao, Mark P Jedrychowski, Devin K Schweppe, Edward L Huttlin, Qing Yu, David E Heppner, Jiaming Li, Jiani Long, Evanna L Mills, John Szpyt, Zhixiang He, Guangyan Du, Ryan Garrity, Anita Reddy, Laura Pontano Vaites, Joao A Paulo, Tinghu Zhang, Nathanael S Gray, Steven P Gygi, Edward T Chouchani
ABSTRACT
Mammalian tissues engage in specialized physiology that is regulated through reversible modification of protein cysteine residues by reactive oxygen species (ROS). ROS regulate a myriad of biological processes, but the protein targets of ROS modification that drive tissue-specific physiology in vivo are largely unknown. Here, we develop Oximouse, a comprehensive and quantitative mapping of the mouse cysteine redox proteome in vivo. We use Oximouse to establish several paradigms of physiological redox signaling. We define and validate cysteine redox networks within each tissue that are tissue selective and underlie tissue-specific biology. We describe a common mechanism for encoding cysteine redox sensitivity by electrostatic gating. Moreover, we comprehensively identify redox-modified disease networks that remodel in aged mice, establishing a systemic molecular basis for the long-standing proposed links between redox dysregulation and tissue aging. We provide the Oximouse compendium as a framework for understanding mechanisms of redox regulation in physiology and aging.
Product Description
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in glass vials
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Insulin from bovine pancreas, ≥25 USP units/mg (HPLC), powder
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine, ≥95% (HPLC), powder
Sigma-Aldrich
Iodoacetic acid N-hydroxysuccinimide ester, powder
Sigma-Aldrich
3-Aminopropylphosphonic acid, 98%
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
2-Aminoethylphosphonic acid, 99%
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Diamide
Sigma-Aldrich
Triethylammonium bicarbonate buffer, 1.0 M, pH 8.5±0.1
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
6-Aminohexylphosphonic acid hydrochloride salt, ≥95%