Merck
  • Caspase-dependent cell death mediates the early phase of aortic hypertrophy regression in losartan-treated spontaneously hypertensive rats.
Caspase-dependent cell death mediates the early phase of aortic hypertrophy regression in losartan-treated spontaneously hypertensive rats.
Circulation research (2003-03-08)
Eve-Lyne Marchand, Shant Der Sarkissian, Pavel Hamet, Denis deBlois
ABSTRACT
Blockade of angiotensin type 1 (AT1) receptors induces smooth muscle cell (SMC) death and regression of aortic hypertrophy in spontaneously hypertensive rats (SHR). We postulated that SMC death and vascular remodeling in this model may be attenuated by z-Val-Ala-Asp(OMe)-CH2F (z-VAD-fmk), a tripeptide inhibitor of caspase enzymes mediating apoptosis. To determine the time course of SMC death and aortic remodeling, SHR were treated with losartan (30 mg/kg per day) for up to 9.5 days. Transient SMC apoptosis occurred in the aortic media with a peak around day 5 of treatment, with increases in the Bax to Bcl-2 protein ratio (>3-fold), in active caspase-3 (5.6-fold), in TUNEL-positive nuclei (19-fold), preceding by 24 hours the peak activation of capase-9 (3.8-fold), and significant reductions in SMC number (46%) and aortic cross-sectional area (8.5%) at 5.5 days. The decrease in total aortic DNA reached significance at 6.5 days (29%). Blood pressure reduction with losartan was progressive and reached significance at day 7 of treatment. Next, we examined the causal link between vascular apoptosis and remodeling. SHR received placebo or losartan (30 mg/kg per day) for 6 days. During the last 24 hours, a subgroup of losartan-treated rats received 3 IV injections of z-VAD-fmk (cumulative dose: 4.4 mg x kg(-1)). All other rats received the vehicle, DMSO. The 24-hour cotreatment with z-VAD-fmk effectively prevented losartan-induced caspase-3 activation and internucleosomal DNA fragmentation, as well as SMC depletion and the reductions in aortic mass and DNA content. Together, these data suggest that caspase-dependent SMC death mediates the early phase of vascular remodeling in response to AT1 receptor blockade in this model of hypertension.
Sigma-Aldrich
Caspase 1 Substrate
Sigma-Aldrich
Caspase 1 Substrate
Sigma-Aldrich
Caspase1 Substrate
Sigma-Aldrich
Caspase 1 Substrate
Sigma-Aldrich
Caspase 1 Substrate
Sigma-Aldrich
Caspase 1 Substrate (ICE), fluorogenic
Sigma-Aldrich
Caspase 1 Substrate
Sigma-Aldrich
Caspase 1 Substrate
Sigma-Aldrich
Caspase 1 Substrate, ≥92% (HPLC)