Merck
  • A peptide inhibitor of Tau-SH3 interactions ameliorates amyloid-β toxicity.
A peptide inhibitor of Tau-SH3 interactions ameliorates amyloid-β toxicity.
Neurobiology of disease (2019-11-08)
Travis Rush, Jonathan R Roth, Samantha J Thompson, Adam R Aldaher, J Nicholas Cochran, Erik D Roberson
ABSTRACT
The microtubule-associated protein Tau is strongly implicated in Alzheimer's disease (AD) and aggregates into neurofibrillary tangles in AD. Genetic reduction of Tau is protective in several animal models of AD and cell culture models of amyloid-β (Aβ) toxicity, making it an exciting therapeutic target for treating AD. A variety of evidence indicates that Tau's interactions with Fyn kinase and other SH3 domain-containing proteins, which bind to PxxP motifs in Tau's proline-rich domain, may contribute to AD deficits and Aβ toxicity. Thus, we sought to determine if inhibiting Tau-SH3 interactions ameliorates Aβ toxicity. We developed a peptide inhibitor of Tau-SH3 interactions and a proximity ligation assay (PLA)-based target engagement assay. Then, we used membrane trafficking and neurite degeneration assays to determine if inhibiting Tau-SH3 interactions ameliorated Aβ oligomer (Aβo)-induced toxicity in primary hippocampal neurons from rats. We verified that Tau reduction ameliorated Aβo toxicity in neurons. Using PLA, we identified a peptide inhibitor that reduced Tau-SH3 interactions in HEK-293 cells and primary neurons. This peptide reduced Tau phosphorylation by Fyn without affecting Fyn's kinase activity state. In primary neurons, endogenous Tau-Fyn interaction was present primarily in neurites and was reduced by the peptide inhibitor, from which we inferred target engagement. Reducing Tau-SH3 interactions in neurons ameliorated Aβo toxicity by multiple outcome measures, namely Aβo-induced membrane trafficking abnormalities and neurite degeneration. Our results indicate that Tau-SH3 interactions are critical for Aβo toxicity and that inhibiting them is a promising therapeutic target for AD.
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Supelco
1,1,1,3,3,3-Hexafluoro-2-propanol, for GC derivatization, LiChropur, ≥99.8%
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline