Merck
  • Increased chemoresistance via Snail-Raf kinase inhibitor protein signaling in colorectal cancer in response to a nicotine derivative.
Increased chemoresistance via Snail-Raf kinase inhibitor protein signaling in colorectal cancer in response to a nicotine derivative.
Oncotarget (2016-03-19)
Tsai-Yu Lee, Chia-Lin Liu, Yun-Ching Chang, Shin Nieh, Yaoh-Shiang Lin, Shu-Wen Jao, Su-Feng Chen, Tsung-Yun Liu
ABSTRACT
A tobacco-specific component, 4-methylnitrosamino-1-3-pyridyl-1-butanone (NNK), is a major risk factor for many cancers. Recent reports have demonstrated that NNK exposure may be associated with tumor progression and chemoresistance in certain cancers. However, the underlying NNK-induced mechanism contributing to the aggressiveness of colorectal cancer (CRC) has not been thoroughly studied. In this study, we used HT29 cells treated with NNK to simulate the long-term exposure of cigarette smoke. A comparative analysis was performed to evaluate cell proliferation, migration, and invasion as well as epithelial-mesenchymal transition (EMT) markers and drug-resistance genes expression, cancer stem cell (CSC) properties, and anti-apoptotic activity. Signaling pathways related to chemoresistance were also investigated. As a result, NNK exposure dose-dependently stimulates cell proliferation, enhance abilities of migration and invasion, induce EMT phenomenon, and attenuate apoptosis. Furthermore, NNK exposure also promotes the capabilities of sphere formation, upregulation of Snail, and overexpression of CD133, Nanog, OCT4, and the drug-resistant genes. Knockdown of Snail results in upregulation of Raf kinase inhibitor protein (RKIP), increased apoptosis, reversal of EMT phenomenon, and reducation of expression of CSC markers, all of which contribute to a decrease of chemoresistance. Our study demonstrates a number of related mechanisms that mediate the effect of NNK exposure on increasing CRC therapeutic resistance via the Snail signaling pathway. Targeting Snail may provide a feasible strategy for the treatment of CRC.
Sigma-Aldrich
Anti-BCRP1 Antibody, clone 5D3, clone 5D3, Chemicon®, from mouse
Sigma-Aldrich
Anti-MDR1 Antibody, extracellular human specific Pgp, clone MC57, clone MC57, Chemicon®, from mouse