Merck
  • Effect of Lycium bararum polysaccharides on methylmercury-induced abnormal differentiation of hippocampal stem cells.
Effect of Lycium bararum polysaccharides on methylmercury-induced abnormal differentiation of hippocampal stem cells.
Experimental and therapeutic medicine (2016-07-23)
Jian-Ying Tian, Wei-Wei Chen, Jing Cui, Hao Wang, Ci Chao, Zhi-Yan Lu, Yong-Yi Bi
ABSTRACT
The aim of the present study was to observe the effects of a general extract of Lycium bararum polysaccharides (LBPs) on methylmercury (MeHg)-induced damage in hippocampus neural stem cells (hNSCs). The hippocampal tissues of embryonic day 16 Sprague-Dawley rats were extracted for the isolation, purification and cloning of hNSCs. Following passage and proliferation for 10 days, the cells were allocated at random into the following groups: Control, LBPs, MeHg and MeHg + LBPs. MTT and microtubule-associated protein 2 (MAP-2)/glial fibrillary acidic protein/Hoechst immunofluorescence tests were performed to detect the differentiation and growth of hNSCs in the various groups. The differentiation rate of MeHg-treated hNSCs and the perimeter of MAP-2-positive neurons were 3.632±0.63% and 62.36±5.58 µm, respectively, significantly lower compared with the control group values of 6.500±0.81% and 166±8.16 µm (P<0.05). Furthermore, the differentiation rate and the perimeter of MAP-2-positive neurons in LBPs groups cells was 7.75±0.59% and 253.3±11.21 µm, respectively, significantly higher compared with the control group (P<0.05). The same parameters in the MeHg + LBPs group were 5.92±0.98% and 111.9±6.07 µm, respectively, significantly higher than the MeHg group (P<0.05). The astrocyte differentiation rates in the MeHg and MeHg + LBPs group were 41.19±2.14 and 34.58±1.70, respectively (P<0.05). These results suggest that LBPs may promote the generation and development of new neurons and inhibit the MeHg-induced abnormal differentiation of astrocytes. Thus, LBPs may be considered to be a potential new treatment for MeHg-induced neurotoxicity in hNSCs.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Cy3 conjugate, Chemicon®, from goat
Sigma-Aldrich
Anti-BrdU Antibody, clone 131-14871, clone 131-14871, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, Chemicon®, from mouse
Sigma-Aldrich
Anti-Nestin Antibody, clone rat-401, clone rat-401, Chemicon®, from mouse
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, FITC conjugate, Species Adsorbed, 1.3 mg/mL, Chemicon®
Sigma-Aldrich
Anti-MAP-2 Antibody, from rabbit, purified by affinity chromatography