Merck
  • Uremic metabolites impair skeletal muscle mitochondrial energetics through disruption of the electron transport system and matrix dehydrogenase activity.
Uremic metabolites impair skeletal muscle mitochondrial energetics through disruption of the electron transport system and matrix dehydrogenase activity.
American journal of physiology. Cell physiology (2019-07-11)
Trace Thome, Zachary R Salyers, Ravi A Kumar, Dongwoo Hahn, Fabian N Berru, Leonardo F Ferreira, Salvatore T Scali, Terence E Ryan
ABSTRACT
Chronic kidney disease (CKD) leads to increased skeletal muscle fatigue, weakness, and atrophy. Previous work has implicated mitochondria within the skeletal muscle as a mediator of muscle dysfunction in CKD; however, the mechanisms underlying mitochondrial dysfunction in CKD are not entirely known. The purpose of this study was to define the impact of uremic metabolites on mitochondrial energetics. Skeletal muscle mitochondria were isolated from C57BL/6N mice and exposed to vehicle (DMSO) or varying concentrations of uremic metabolites: indoxyl sulfate, indole-3-acetic-acid, l-kynurenine, and kynurenic acid. A comprehensive mitochondrial phenotyping platform that included assessments of mitochondrial oxidative phosphorylation (OXPHOS) conductance and respiratory capacity, hydrogen peroxide production (JH2O2), matrix dehydrogenase activity, electron transport system enzyme activity, and ATP synthase activity was employed. Uremic metabolite exposure resulted in a ~25-40% decrease in OXPHOS conductance across multiple substrate conditions (P < 0.05, n = 5-6/condition), as well as decreased ADP-stimulated and uncoupled respiratory capacity. ATP synthase activity was not impacted by uremic metabolites; however, a screen of matrix dehydrogenases indicated that malate and glutamate dehydrogenases were impaired by some, but not all, uremic metabolites. Assessments of electron transport system enzymes indicated that uremic metabolites significantly impair complex III and IV. Uremic metabolites resulted in increased JH2O2 under glutamate/malate, pyruvate/malate, and succinate conditions across multiple levels of energy demand (all P < 0.05, n = 4/group). Disruption of mitochondrial OXPHOS was confirmed by decreased respiratory capacity and elevated superoxide production in cultured myotubes. These findings provide direct evidence that uremic metabolites negatively impact skeletal muscle mitochondrial energetics, resulting in decreased energy transfer, impaired complex III and IV enzyme activity, and elevated oxidant production.
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
Sigma-Aldrich
Myxothiazol, from Myxococcus fulvus Mx f85, ≥98% (HPLC)
Sigma-Aldrich
Sodium cacodylate Buffer pH 6.5; 50 mM – Isopropanol 15% solution
Sigma-Aldrich
Malonic acid, ReagentPlus®, 99%
Sigma-Aldrich
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, ≥97.0% (AT)
Sigma-Aldrich
Potassium cyanide, BioUltra, ≥98.0% (AT)
Sigma-Aldrich
3-Methyl-2-oxopentanoic acid sodium salt, ≥98%
Sigma-Aldrich
α-Ketoglutaric acid, ≥98.5% (NaOH, titration)
Sigma-Aldrich
Adenosine 5′-diphosphate monopotassium salt dihydrate, bacterial, ≥95%, powder
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide sodium salt
Sigma-Aldrich
n-Dodecyl β-D-maltoside, ≥98% (GC)
Sigma-Aldrich
Creatine Phosphokinase from rabbit muscle, Type I, salt-free, lyophilized powder, ≥150 units/mg protein
Sigma-Aldrich
Phosphocreatine di(tris) salt, ≥97% (enzymatic)
Sigma-Aldrich
Peroxidase from horseradish, Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Auranofin, ≥98% (HPLC)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Kynurenic acid, ≥98%
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
(±)-3-Methyl-2-oxovaleric acid sodium salt
Sigma-Aldrich
Decylubiquinone, ≥97% (HPLC)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Roche
Phosphoenol-pyruvate, 97% (PEP-K), pkg of 1 g
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Ethidium homodimer, suitable for fluorescence, ~90% (HPCE)
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
L-Kynurenine, ≥98% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MES potassium salt, ≥99% (titration)