Merck
  • Spatial oxidation of L-plastin downmodulates actin-based functions of tumor cells.
Spatial oxidation of L-plastin downmodulates actin-based functions of tumor cells.
Nature communications (2019-09-11)
Emre Balta, Robert Hardt, Jie Liang, Henning Kirchgessner, Christian Orlik, Beate Jahraus, Stefan Hillmer, Stefan Meuer, Katrin Hübner, Guido H Wabnitz, Yvonne Samstag
ABSTRACT
Several antitumor therapies work by increasing reactive oxygen species (ROS) within the tumor micromilieu. Here, we reveal that L-plastin (LPL), an established tumor marker, is reversibly regulated by ROS-induced thiol oxidation on Cys101, which forms a disulfide bridge with Cys42. LPL reduction is mediated by the Thioredoxin1 (TRX1) system, as shown by TRX1 trapping, TRX1 knockdown and blockade of Thioredoxin1 reductase (TRXR1) with auranofin. LPL oxidation diminishes its actin-bundling capacity. Ratiometric imaging using an LPL-roGFP-Orp1 fusion protein and a dimedone-based proximity ligation assay (PLA) reveal that LPL oxidation occurs primarily in actin-based cellular extrusions and strongly inhibits cell spreading and filopodial extension formation in tumor cells. This effect is accompanied by decreased tumor cell migration, invasion and extracellular matrix (ECM) degradation. Since LPL oxidation occurs following treatment of tumors with auranofin or γ-irradiation, it may be a molecular mechanism contributing to the effectiveness of tumor treatment with redox-altering therapies.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Swagelok® Miniature Quick Connect Body Assembly, Swagelok® QM2-B-200, 1/8 in. Swagelok fitting, stainless steel
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein