Merck
  • IκBβ-mediated NF-κB activation confers protection against hyperoxic lung injury.
IκBβ-mediated NF-κB activation confers protection against hyperoxic lung injury.
American journal of respiratory cell and molecular biology (2013-09-27)
Katherine A Michaelis, Fadeke Agboke, Thanh Liu, Kristie Han, Manasa Muthu, Csaba Galambos, Guang Yang, Phyllis A Dennery, Clyde J Wright
ABSTRACT
Supplemental oxygen is frequently used in an attempt to improve oxygen delivery; however, prolonged exposure results in damage to the pulmonary endothelium and epithelium. Although NF-κB has been identified as a redox-responsive transcription factor, whether NF-κB activation exacerbates or attenuates hyperoxic lung injury is unclear. We determined that sustained NF-κB activity mediated by IκBβ attenuates lung injury and prevents mortality in adult mice exposed to greater than 95% O2. Adult wild-type mice demonstrated evidence of alveolar protein leak and 100% mortality by 6 days of hyperoxic exposure, and showed NF-κB nuclear translocation that terminated after 48 hours. Furthermore, these mice showed increased expression of NF-κB-regulated proinflammatory and proapoptotic cytokines. In contrast, mice overexpressing the NF-κB inhibitory protein, IκBβ (AKBI), demonstrated significant resistance to hyperoxic lung injury, with 50% surviving through 8 days of exposure. This was associated with NF-κB nuclear translocation that persisted through 96 hours of exposure. Although induction of NF-κB-regulated proinflammatory cytokines was not different between wild-type and AKBI mice, significant up-regulation of antiapoptotic proteins (BCL-2, BCL-XL) was found exclusively in AKBI mice. We conclude that sustained NF-κB activity mediated by IκBβ protects against hyperoxic lung injury through increased expression of antiapoptotic genes.
Sigma-Aldrich
Anti-α-Tubulin Antibody, clone DM1A, clone DM1A, Upstate®, from mouse
Sigma-Aldrich
Mouse IL-11 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Anti-Mn-SOD Antibody, Upstate®, from rabbit