Merck
  • Cerium Oxide Nanoparticles Decrease Drought-Induced Oxidative Damage in Sorghum Leading to Higher Photosynthesis and Grain Yield.
Cerium Oxide Nanoparticles Decrease Drought-Induced Oxidative Damage in Sorghum Leading to Higher Photosynthesis and Grain Yield.
ACS omega (2018-11-10)
Maduraimuthu Djanaguiraman, Remya Nair, Juan Pablo Giraldo, Pagadala Venkata Vara Prasad
ABSTRACT
Drought is a major abiotic stress affecting crop growth and yield worldwide. Drought-induced oxidative stress results in the reduction of plant photosynthesis and reproductive success. Cerium oxide nanoparticles (nanoceria) possess potent antioxidant properties that can alleviate drought-induced oxidative stress by catalytic scavenging reactive oxygen species (ROS), thereby protecting sorghum [Sorghum bicolor (L.) Moench] photosynthesis and grain yield. Drought was imposed at the booting stage by withholding water for 21 d. Foliar-sprayed nanoceria (10 mg L-1) efficiently reduced leaf superoxide radical (41%) and hydrogen peroxide (36%) levels and decreased cell membrane lipid peroxidation (37%) under drought. Nanoceria increased leaf carbon assimilation rates (38%), pollen germination (31%), and seed yield per plant (31%) in drought-stressed plants relative to water-sprayed controls. Translocation study indicated that nanoceria can move from root to shoot of sorghum plants. Toxicity assays in mammalian cells indicated that nanoceria effective concentration (EC)50 of >250 mg L-1 is well above the concentration used in this study. Foliar-sprayed nanoceria protect sorghum plants from oxidative damage under drought stress leading to higher grain yield.
Sigma-Aldrich
BrdU (5-Bromo-2′-deoxyuridine), Used for the study of DNA synthesis where it is incorporated into DNA in place of thymidine, BrdU can be used in conjunction with anti-BrdU for immunocytochemical analysis of cell proliferation.