Merck
Identification of the Human Skeletal Stem Cell.
Cell (2018-09-23)
Charles K F Chan, Gunsagar S Gulati, Rahul Sinha, Justin Vincent Tompkins, Michael Lopez, Ava C Carter, Ryan C Ransom, Andreas Reinisch, Taylor Wearda, Matthew Murphy, Rachel E Brewer, Lauren S Koepke, Owen Marecic, Anoop Manjunath, Eun Young Seo, Tripp Leavitt, Wan-Jin Lu, Allison Nguyen, Stephanie D Conley, Ankit Salhotra, Thomas H Ambrosi, Mimi R Borrelli, Taylor Siebel, Karen Chan, Katharina Schallmoser, Jun Seita, Debashis Sahoo, Henry Goodnough, Julius Bishop, Michael Gardner, Ravindra Majeti, Derrick C Wan, Stuart Goodman, Irving L Weissman, Howard Y Chang, Michael T Longaker
ABSTRACT
Stem cell regulation and hierarchical organization of human skeletal progenitors remain largely unexplored. Here, we report the isolation of a self-renewing and multipotent human skeletal stem cell (hSSC) that generates progenitors of bone, cartilage, and stroma, but not fat. Self-renewing and multipotent hSSCs are present in fetal and adult bones and can also be derived from BMP2-treated human adipose stroma (B-HAS) and induced pluripotent stem cells (iPSCs). Gene expression analysis of individual hSSCs reveals overall similarity between hSSCs obtained from different sources and partially explains skewed differentiation toward cartilage in fetal and iPSC-derived hSSCs. hSSCs undergo local expansion in response to acute skeletal injury. In addition, hSSC-derived stroma can maintain human hematopoietic stem cells (hHSCs) in serum-free culture conditions. Finally, we combine gene expression and epigenetic data of mouse skeletal stem cells (mSSCs) and hSSCs to identify evolutionarily conserved and divergent pathways driving SSC-mediated skeletogenesis. VIDEO ABSTRACT.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
N-Chlorosuccinimide, 98%
Sigma-Aldrich
Oil Red O, certified by the Biological Stain Commission
Sigma-Aldrich
Phosphotungstic acid hydrate, reagent grade
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Tris Buffered Saline, 10 ×, solution
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Anti-CD164 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Histopaque®-1119, sterile-filtered, density: 1.119
Sigma-Aldrich
Anti-Perilipin A Antibody, Chemicon®, from rabbit