Merck
  • An Evaluation of the in vivo Safety of Nonporous Silica Nanoparticles: Ocular Topical Administration versus Oral Administration.
An Evaluation of the in vivo Safety of Nonporous Silica Nanoparticles: Ocular Topical Administration versus Oral Administration.
Scientific reports (2017-08-16)
Martha Kim, Joo-Hee Park, Hyejoong Jeong, Jinkee Hong, Woo Sung Choi, Byung-Han Lee, Choul Yong Park
ABSTRACT
Nonporous silica nanoparticles (SiNPs) have potential as promising carriers for ophthalmic drugs. However, the in vivo safety of ocular topical SiNPs remains unclear. This study investigated the in vivo safety of oral and ocular topical applications of 100 nm-sized SiNPs in Sprague-Dawley rats. The rats were divided into the following four groups: low-dose oral administration (total 100 mg/kg of SiNPs mixed with food for one week), high-dose oral administration (total 1000 mg/kg of SiNPs mixed with food for one week), ocular topical administration (10 mg/ml concentration, one drop, applied to the right eyes four times a day for one month), or a negative control (no SiNP treatment). The rats were observed for 12 weeks to investigate any signs of general or ocular toxicity. During the observation period, no differences were observed in the body weights, food and water intakes, behaviors and abnormal symptoms of the four groups. No animal deaths occurred. After 12 weeks, hematologic, blood biochemical parameters and ophthalmic examinations revealed no abnormal findings in any of the animals. The lack of toxicity of the SiNPs was further verified in autopsy findings of brain, liver, lung, spleen, heart, kidneys, intestine, eyeballs, and ovaries or testes.
Sigma-Aldrich
Silica, monodisperse, non-porous, 50 μm
Sigma-Aldrich
Silica, monodisperse, non-porous, 200 nm
Sigma-Aldrich
Silica, monodisperse, non-porous, 750 nm
Sigma-Aldrich
Silica, monodisperse, non-porous, 150 nm
Sigma-Aldrich
Silica, monodisperse, non-porous, 300 nm
Sigma-Aldrich
Silica, monodisperse, non-porous, 3.0 μm
Sigma-Aldrich
Silica, monodisperse, non-porous, 400 nm
Sigma-Aldrich
Silica, monodisperse, non-porous, 2.0 μm
Sigma-Aldrich
Silica, monodisperse, non-porous, 1.0 μm
Sigma-Aldrich
Silica, monodisperse, non-porous, 10 μm
Sigma-Aldrich
Silica, monodisperse, non-porous, 500 nm
Sigma-Aldrich
Silica, monodisperse, non-porous, 20 μm
Sigma-Aldrich
Silica, monodisperse, non-porous, 70 μm
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 900 nm particle size, 5 % (w/w) in water, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 900 nm particle size, 5 % (w/w) in ethanol, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 300 nm particle size, 5 % (w/w) in ethanol, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 500 nm particle size, 5 % (w/w) in water, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 700 nm particle size, 5 % (w/w) in water, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 700 nm particle size, 5 % (w/w) in ethanol, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 500 nm particle size, 5 % (w/w) in ethanol, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 150 nm particle size, 5 % (w/w) in ethanol, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 300 nm particle size, 5 % (w/w) in water, dispersion
Sigma-Aldrich
Silica nanoparticles, pore size non-porous, 150 nm particle size, 5 % (w/w) in water, dispersion