Merck
  • Genome-scale CRISPR-Cas9 knockout and transcriptional activation screening.
Genome-scale CRISPR-Cas9 knockout and transcriptional activation screening.
Nature protocols (2017-03-24)
Julia Joung, Silvana Konermann, Jonathan S Gootenberg, Omar O Abudayyeh, Randall J Platt, Mark D Brigham, Neville E Sanjana, Feng Zhang
ABSTRACT
Forward genetic screens are powerful tools for the unbiased discovery and functional characterization of specific genetic elements associated with a phenotype of interest. Recently, the RNA-guided endonuclease Cas9 from the microbial CRISPR (clustered regularly interspaced short palindromic repeats) immune system has been adapted for genome-scale screening by combining Cas9 with pooled guide RNA libraries. Here we describe a protocol for genome-scale knockout and transcriptional activation screening using the CRISPR-Cas9 system. Custom- or ready-made guide RNA libraries are constructed and packaged into lentiviral vectors for delivery into cells for screening. As each screen is unique, we provide guidelines for determining screening parameters and maintaining sufficient coverage. To validate candidate genes identified by the screen, we further describe strategies for confirming the screening phenotype, as well as genetic perturbation, through analysis of indel rate and transcriptional activation. Beginning with library design, a genome-scale screen can be completed in 9-15 weeks, followed by 4-5 weeks of validation.
Sigma-Aldrich
Mouse Whole Genome SAM CRISPRa Pooled Lentiviral Library Kit Puro
Sigma-Aldrich
dCas9-VP64-Blasticidin SAM CRISPRa Helper Construct 1 Plasmid DNA
Sigma-Aldrich
dCas9-VP64-Blasticidin SAM CRISPRa Helper Construct 1 Lentiviral Transduction Particles
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Tris-EDTA buffer solution, BioUltra, for molecular biology, pH 8.0
Sigma-Aldrich
ROCK Inhibitor (Y-27632), The ROCK Inhibitor (Y-27632) is available in a 5 mg format & has been optimized & validated for cell culture & Neuroscience applications.
Sigma-Aldrich
MS2-P65-HSF1-Hygromycin SAM CRISPRa Helper Construct 2 Lentiviral Transduction Particles
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Proteinase K Inhibitor, Proteinase K Inhibitor is a tetrapeptidyl chloromethyl ketone compound that acts as an active-site-targeting irreversible inhibitor against proteinase K.
Sigma-Aldrich
Human Whole Genome SAM CRISPRa Pooled Lentiviral Library Kit Puro
Sigma-Aldrich
Human Whole Genome SAM CRISPRa Pooled Lentiviral Library Kit Zeo
Sigma-Aldrich
Mouse Whole Genome SAM CRISPRa Pooled Lentiviral Library Kit Zeo
Sigma-Aldrich
SAM CRISPRa Helper Construct Kit Lentiviral Transduction Particles
Sigma-Aldrich
MS2-P65-HSF1-Hygromycin SAM CRISPRa Helper Construct 2 Plasmid DNA
Sigma-Aldrich
SAM CRISPRa Helper Construct Kit Plasmid DNA