Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Yttrium(III) oxide
Synonym(s):
Yttria
Linear Formula:
Y
2
O
3
CAS Number:
1314-36-9
Molecular Weight:
225.81
EC Number:
215-233-5
Products
Brand
Sigma-Aldrich (3)
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
205168
Yttrium(III) oxide,
99.99% trace metals basis
Expand
View Pricing
544892
Yttrium(III) oxide,
nanopowder, <50 nm particle size
Expand
View Pricing
204927
Yttrium(III) oxide,
99.999% trace metals basis
Expand
View Pricing