Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Xanthine Oxidase from bovine milk
Synonym(s):
XOD, Xanthine:oxygen oxidoreductase
CAS Number:
9002-17-9
EC Number:
232-657-6
Products
Product Category
enzymes (3)
Brand
Sigma-Aldrich (4)
Biological Source
Color
Physical Form
Tissue
Chemical Composition
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
X1875
Xanthine Oxidase from bovine milk,
Grade I, ammonium sulfate suspension, ≥0.4 units/mg protein
Expand
View Pricing
X4376
Xanthine Oxidase from bovine milk,
lyophilized powder, 0.4-1.0 units/mg protein
Expand
View Pricing
X4875
Xanthine Oxidase from bovine milk,
Grade IV, ammonium sulfate suspension, ≥0.1 units/mg protein
Expand
View Pricing
X4500
Xanthine Oxidase from bovine milk,
Grade III, ammonium sulfate suspension, ≥0.8 units/mg protein
Expand
View Pricing