Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Worthington Accessories
Products
Product Category
racks (4)
Brand
SAFC (1)
Sigma-Aldrich (1)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
R0509
Worthington Accessories,
cast aluminum roller base with 5 wheels for 35 L Dewar
Expand
View Pricing
R0384
Worthington Accessories,
5 shelf rack for LS750 Dewar
Expand
View Pricing
C0544
Worthington Accessories,
cap/necktube core for 35 L Dewar
Expand
View Pricing
S8777
Worthington Accessories,
tipping stand for 25 L Dewar
Expand
View Pricing
Z744002
Worthington Accessories,
rack for 8 drawers for LS4800 liquid nitrogen refrigerator
Expand
View Pricing
M9904
Worthington Accessories,
measuring rod for 35 L Dewar
Expand
View Pricing
C0796
Worthington Accessories,
cap/necktube core for 10 L Dewar
Expand
View Pricing
Z744004
Worthington Accessories,
rack for 5 drawers for LS3000 liquid nitrogen refridgerator
Expand
View Pricing
Z743999
Worthington Accessories,
necktube core for LS6000 liquid nitrogen refrigerator
Expand
View Pricing
Z744003
Worthington Accessories,
necktube core for LS3000 liquid nitrogen refrigerator
Expand
View Pricing
C7431
Worthington Accessories,
cap/necktube core for 25 L Dewar
Expand
View Pricing
Z744000
Worthington Accessories,
rack for 10 drawers for LS6000 liquid nitrogen refrigerator
Expand
View Pricing
C0671
Worthington Accessories,
cap for 4 L Dewar
Expand
View Pricing
Z744005
Worthington Accessories,
necktube core for LD35 and LD50 dewars
Expand
View Pricing
Z744001
Worthington Accessories,
necktube core for LS4800 liquid nitrogen refrigerator
Expand
View Pricing