Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
Benchkote
®
surface protector
Products
Manufacturer
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
WHA2300916
Whatman
®
Benchkote
®
surface protector,
Benchkote surface protector, sheets, 460 x 570mm, 50/pk
Expand
View Pricing
WHA2300731
Whatman
®
Benchkote
®
surface protector,
Benchkote surface protector, reel, 460 mm x 50 m, 1/pk
Expand
View Pricing
WHA2300917
Whatman
®
Benchkote
®
surface protector,
Benchkote surface protector, sheets, 460 x 570 mm, 100/pk
Expand
View Pricing
WHA23016160
Whatman
®
Benchkote
®
surface protector,
Benchkote Plus surface protector, reel, 600 mm x 50 m, 1/pk
Expand
View Pricing
WHA23016150
Whatman
®
Benchkote
®
surface protector,
Benchkote Plus surface protector, sheets, 500 x 600 mm, 50/pk
Expand
View Pricing
WHA2300772
Whatman
®
Benchkote
®
surface protector,
Benchkote surface protector, reel, 920 mm x 50 m, 1/pk
Expand
View Pricing
WHA2300594
Whatman
®
Benchkote
®
surface protector,
Benchkote surface protector, pad, 460 x 570 mm, 50/pk
Expand
View Pricing