Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Viscosity Bath Media
Products
Manufacturer
Format
Application
Reference Material Type
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
BM2
Viscosity Bath Media,
White Oil for use 80 to 120 °C
Expand
View Pricing
BM5
Viscosity Bath Media,
Silicone Fluid 26 cSt at 25 °C for use 120 to 150 °C
Expand
View Pricing
BM1
Viscosity Bath Media,
White Oil for use 40 to 80 °C
Expand
View Pricing
BM6
Viscosity Bath Media,
Silicone Fluid 20 cSt at 25 °C for use 90 to 135 °C
Expand
View Pricing
BM7
Viscosity Bath Media,
Silicone Fluid 10 cSt at 25 °C for use 50 to 100 °C
Expand
View Pricing
BM8
Viscosity Bath Media,
Silicone Fluid 5 cSt at 25 °C for use 20 to 60 °C
Expand
View Pricing