Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vials, crimp top, serum bottle
Products
Product Category
bottles (8)
Brand
Supelco (7)
OD (mm)
Volume (ml)
Closure Type
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
33110-U
Vials, crimp top, serum bottle,
volume 100 mL, clear glass bottle, O.D. 51.7 mm × H (94.5 mm), pkg of 36 ea
Expand
View Pricing
33108-U
Vials, crimp top, serum bottle,
volume 50 mL, clear glass bottle, O.D. × H 43 mm × 73 mm, pkg of 36 ea
Expand
View Pricing
33109-U
Vials, crimp top, serum bottle,
volume 50 mL, clear glass bottle, O.D. × H 43 mm × 73 mm, pkg of 272 ea
Expand
View Pricing
33111-U
Vials, crimp top, serum bottle,
volume 100 mL, clear glass bottle, O.D. × H 51.7 mm ( ) × 94.5 mm, pkg of 144 ea
Expand
View Pricing
33106
Vials, crimp top, serum bottle,
volume 30 mL, clear glass bottle, O.D. × H 36.6 mm × 62.8 mm, pkg of 36 ea
Expand
View Pricing
33143
Vials, crimp top, serum bottle,
volume 10 mL, amber glass bottle, O.D. × H 25 mm × 50 mm, pkg of 36 ea
Expand
View Pricing
33144
Vials, crimp top, serum bottle,
volume 10 mL, amber glass bottle, O.D. × H 25 mm × 50 mm, pkg of 288 ea
Expand
View Pricing
33107
Vials, crimp top, serum bottle,
volume 30 mL, clear glass bottle, O.D. × H 36.6 mm × 62.8 mm, pkg of 288 ea
Expand
View Pricing