Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Universal carousel pipette stand
Products
Color
Manufacturer
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
HS120499
Universal carousel pipette stand,
assorted colors to clear, 8 clear tops plus 8 assorted colored tops + 8 clear bottoms
Expand
View Pricing
HS120480
Universal carousel pipette stand,
blue to green, 8 of each color for top + 8 blue bottoms
Expand
View Pricing
HS120481
Universal carousel pipette stand,
purple to clear, 8 of each color for top + 8 clear bottoms
Expand
View Pricing