Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trypsin inhibitor
Synonym(s):
SBTI, Ovomucoid, Kunitz Inhibitor, Trypsin inhibitor from
Glycine max
(soybean), Trypsin Inhibitor from lima beans
CAS Number:
9035-81-8
EC Number:
232-906-9
Products
Product Category
inhibitors (8)
proteases (1)
Brand
Sigma-Aldrich (9)
Biological Source
Physical Form
Product Line
Tissue
Quality Segment
Application
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
T9128
Trypsin inhibitor,
lyophilized powder, suitable for inhibition assay
Expand
View Pricing
T9253
Trypsin inhibitor,
powder
Expand
View Pricing
T9003
Trypsin inhibitor,
lyophilized powder
Expand
View Pricing
T2327
Trypsin inhibitor,
lyophilized powder, ≥95% (Kunitz inhibitor, SDS-PAGE)
Expand
View Pricing
T2011
Trypsin inhibitor,
powder
Expand
View Pricing
SRE0044
Trypsin inhibitor,
lyophilized powder, suitable for mammalian cell culture
Expand
View Pricing
T9378
Trypsin inhibitor,
crude powder
Expand
View Pricing
T1021
Trypsin inhibitor,
powder, suitable for isoelectric focusing (IEF)
Expand
View Pricing
T9767
Trypsin inhibitor,
powder, suitable for electrophoresis
Expand
View Pricing