Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TrueNorth
®
Cool Container with Lid
Products
Capacity (L)
Diameter (mm)
Height (mm)
Color
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
HS28721G
TrueNorth
®
Cool Container with Lid,
Ice Pan, capacity 1 L, green (polyurethane)
Expand
View Pricing
HS28724B
TrueNorth
®
Cool Container with Lid,
Ice Pan, capacity 4 L, blue (polyurethane)
Expand
View Pricing
HS28714K
TrueNorth
®
Cool Container with Lid,
Ice Bucket, capacity 4 L, black (polyurethane)
Expand
View Pricing
HS28729R
TrueNorth
®
Cool Container with Lid,
Ice Pan, capacity 9 L, red (polyurethane)
Expand
View Pricing