Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trizma
®
Pre-set crystals
Synonym(s):
Tris crystals
Products
Product Category
buffers (18)
Brand
Sigma-Aldrich (17)
Feature
pH Value
Physical Form
Quality Segment
Application
Special Grade
BioPerformance Certified (16)
Technique
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
T7193
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 7.0, average M
w
154.8
Expand
View Pricing
T8443
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 8.0, average M
w
141.8
Expand
View Pricing
T7943
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 7.6, average M
w
149.0
Expand
View Pricing
T7693
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 7.4, average M
w
151.6
Expand
View Pricing
T1878
Trizma
®
Pre-set crystals,
Type 8.3-FT, pH 8.3, suitable for use with marine species
Expand
View Pricing
T8818
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 8.5, average M
w
131.4
Expand
View Pricing
T7818
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 7.5, average M
w
150.6
Expand
View Pricing
T8943
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 8.3, average M
w
135.4
Expand
View Pricing
T9693
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 9.0, average M
w
124.6
Expand
View Pricing
T8568
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 8.1, average M
w
139.8
Expand
View Pricing
T8068
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 7.7, average M
w
147.6
Expand
View Pricing
T8193
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 7.8, average M
w
145.8
Expand
View Pricing
T7443
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 7.2, average M
w
153.8
Expand
View Pricing
T0319
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 7.4, average M
w
151.6
Expand
View Pricing
T9568
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 8.9, average M
w
125.6
Expand
View Pricing
T1194
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 8.5, average M
w
131.4
Expand
View Pricing
T1753
Trizma
®
Pre-set crystals,
Type 7.4-FT, pH 7.4, suitable for use with fresh water species
Expand
View Pricing
T9943
Trizma
®
Pre-set crystals,
BioPerformance Certified, pH 7.0, average M
w
154.8
Expand
View Pricing