Skip to Content
Merck
Triethyl(trifluoromethyl)silane 98%

Triethyl(trifluoromethyl)silane

Triethyl(trifluoromethyl)silane 98%
Synonym(s):
(Trifluoromethyl)triethylsilane, Triethylsilyl trifluoromethane
Linear Formula:
(C2H5)3SiCF3
CAS Number:
Molecular Weight:
184.27

Products

Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing