Skip to Content
Merck
trans-2-Methyl-2-pentenoic acid 97%

trans-2-Methyl-2-pentenoic acid

trans-2-Methyl-2-pentenoic acid 97%
Linear Formula:
C2H5CH=C(CH3)CO2H
CAS Number:
Molecular Weight:
114.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing